Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCRL2 Rat anti-Mouse, PE, Clone: BZ2E3, BD

Catalog No. BDB564946 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB564946 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB564946 Supplier BD Biosciences Supplier No. 564946
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The BZ2E3 monoclonal antibody specifically binds to CCRL2 (C-C chemokine receptor-like 2) which is also known as Lipopolysaccharide-inducible C-C chemokine receptor (L-CCR/Lccr). CCRL2 is a chemokine receptor-like, 7-transmembrane protein that can bind CCL19 or the nonchemokine chemoattractant, chemerin/RARRES2. CCRL2 is expressed by a variety of cell types including mast cells, macrophages and some endothelial cells. CCRL2 appears to play various roles in regulating chemokine activities, e.g., by binding and internalizing excess local levels of the CCL19 /MIP-3β chemokine. Alternatively, endothelial cell-expressed CCRL2 can bind and present the chemerin/RARRES2 chemoattractant to lymphoid cells that express the functional signaling receptor, CMKLR1, and promote their adhesion.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CCRL2
Applications Flow Cytometry
Classification Monoclonal
Clone BZ2E3
Concentration 0.2mg/mL
Conjugate PE
Description C-C chemokine receptor-like 2; L-CCR; Lccr; Cmkbr1l2; Ackr5
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Mouse CCRL2 Peptide
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.