Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

chaperonin containing TCP1, subunit 6B (zeta 2), Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89018242 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-018-242 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-018-242 Supplier Abnova Corporation Supplier No. H00010693D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human CCT6B protein.

This gene encodes a molecular chaperone that is a member of the chaperonin containing TCP1 complex (CCT), also known as the TCP1 ring complex (TRiC). This complex consists of two identical stacked rings, each containing eight different proteins. Unfolded polypeptides enter the central cavity of the complex and are folded in an ATP-dependent manner. The complex folds various proteins, including actin and tubulin. Alternate transcriptional splice variants of this gene have been observed but have not been thoroughly characterized. [provided by RefSeq

Sequence: MAAIKAVNSKAEVARAQAALAVNICAARGLQDVLRTNLGPKGTMKMLASGAGDIKLTKDGNVLLDEMQIQHPTASLIAKVATAQDDVTGDGTTSNVLIIGELLKQADLYISEGLHPRIIAEGFEAAKIKALEVLEEVKVTKEMKRKILLDVARTSLQTKVHAELADVLTEVVVDSVLAVRRPGYPIDLFMVEIMEMKHKLGTDTKLIQGLVLDHGARHPDMKKRVEDAFILICNVSLEYEKTEVNSGFFYKTAEEKEKLVKAERKFIEDRVQKIIDLKDKVCAQSNKGFVVINQKGIDPFSLDSLAKHGIVALRRAKRRNMERLSLACGGMAVNSFEDLTVDCLGHAGLVYEYTLGEEKFTFIEECVNPCSVTLLVKGPNKHTLTQVKDAIRDGLRAIKNAIEDGCMVPGAGAIEVAMAEALVTYKNSIKGRARLGVQAFADALLIIPKVLAQNAGYDPQETLVKVQAEHVESKQLVGVDLNTGEPMVAADAGVWDNYCVKKQLLHSCTVIATNILLVDEIMRAGMSSLK

Specifications

Antigen chaperonin containing TCP1, subunit 6B (zeta 2)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human CCT6B protein.
Formulation PBS with no preservative; pH 7.4
Gene CCT6B
Gene Accession No. BC027591.1
Gene Alias CCT-zeta-2/CCTZ-2/Cctz2/TCP-1-zeta-2/TSA303
Gene Symbols CCT6B
Host Species Rabbit
Immunogen CCT6B (AAH27591.1, 1 a.a. ∼ 530 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10693
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.