Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD10 Mouse anti-Human, Brilliant Violet 605, Clone: HI10a, BD

Catalog No. BDB562978 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB562978 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB562978 Supplier BD Biosciences Supplier No. 562978
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The HI10a monoclonal antibody specifically binds to CD10 which is also known as Neutral endopeptidase (NEP), Enkephalinase, Atriopeptidase, and Neprilysin. CD10 is encoded by MME (membrane metallo-endopeptidase). CD10 is a 100 kDa type II transmembrane glycoprotein that has neutral endopeptidase activity and is otherwise known as the Common Acute Lymphoblastic Leukemia Antigen (CALLA). CD10 is expressed on a wide variety of normal and neoplastic cell types. Normal cells expressing CD10 include granulocytes, bone marrow stromal cells, a subset of B-cell progenitors, germinal center B cells and fibroblasts. This cell surface metalloendopeptidase inactivates a number of signaling molecules and serves as a major regulator in the nervous, immune and other systems.

For optimal and reproducible results, BD Horizon Brilliant Stain Buffer should be used anytime two or more BD Horizon Brilliant dyes are used in the same experiment. Fluorescent dye interactions may cause staining artifacts which may affect data interpretation. The BD Horizon Brilliant Stain Buffer was designed to minimize these interactions. More information can be found in the Technical Data Sheet of the BD Horizon Brilliant Stain Buffer (Cat. No. 563794).

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD10
Applications Flow Cytometry
Classification Monoclonal
Clone HI10a
Conjugate Brilliant Violet 605
Description MME; CALLA; EPN; NEP; neprilysin; SFE; atriopeptidase; enkephalinase
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Acute CALLA Leukemia Blast Cells
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.