Learn More
CD10 Mouse anti-Human, PE, Clone: HI10A, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 340921
Description
Clone HI10a reacts with the human form of the 100kDa type II transmembrane glycoprotein, also known as the common acute lymphoblasticleukemia antigen (CALLA). CD10 is expressed on granulocytes, a subset of B-cell progenitors, germinal center cells and fibroblasts. Clone HI10a also cross-reacts with a subset of peripheral blood granulocytes of baboon, and both rhesus and cynomolgus macaque monkeys. The reactivity pattern on granulocytes is similar to that observed with peripheral blood granulocytes from normal human donors.

Specifications
CD10 | |
Monoclonal | |
6μg/mL | |
Phosphate buffered saline with gelatin and 0.1% sodium azide. | |
Acute CALLA Leukemia Blast Cells | |
50 Tests | |
Primary | |
Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed. |
Flow Cytometry | |
HI10a | |
PE | |
Mouse | |
Affinity Purified | |
RUO | |
Human | |
IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.