Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD10 Mouse anti-Human, PE, Clone: HI10A, BD

Catalog No. BDB561002 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
25 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB561002 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB561002 Supplier BD Biosciences Supplier No. 561002
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Clone HI10a reacts with the human form of the 100kDa type II transmembrane glycoprotein, also known as the common acute lymphoblasticleukemia antigen (CALLA). CD10 is expressed on granulocytes, a subset of B-cell progenitors, germinal center cells and fibroblasts. Clone HI10a also cross-reacts with a subset of peripheral blood granulocytes of baboon, and both rhesus and cynomolgus macaque monkeys. The reactivity pattern on granulocytes is similar to that observed with peripheral blood granulocytes from normal human donors.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD10
Applications Flow Cytometry
Classification Monoclonal
Clone HI10a
Conjugate PE
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Acute CALLA Leukemia Blast Cells
Purification Method Affinity Purified
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.