Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD100 (Sema4D) Mouse anti-Human, Alexa Fluor 647, Clone: A8, BD

Catalog No. BDB564873 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB564873 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB564873 Supplier BD Biosciences Supplier No. 564873
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The A8 monoclonal antibody specifically binds to CD100 which is also known as Semaphorin-4D (SEMA4D). CD100 is a 150 kDa type I transmembrane glycoprotein that belongs to the semaphorin family. CD100 is widely expressed in the nervous and immune systems. Although it is expressed on T cells, its expression is significantly upregulated on activated T cells. It is also expressed on NK cells, B cells, and monocytes. CD100 is an adhesion molecule that functions as a signaling molecule which plays a role in lymphocyte activation. CD100 interaction with CD72 enhances B cell signaling and ultimately promotes B cell activation and survival.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD100
Applications Flow Cytometry
Classification Monoclonal
Clone A8
Conjugate Alexa Fluor 647
Description Semaphorin-4D ; SEMA4D; SEM4D; SEMAJ; GR3; C9orf164
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.