Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD100 Mouse, Unlabeled, Clone: 30, BD

Catalog No. BDB610670 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610670 50 μg
BDB610671 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610670 Supplier BD Biosciences Supplier No. 610670
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD100 was originally described as a 150 kDa surface T-cell antigen induced after T-cell activation. Human CD100 exhibits widespread gene expression but is most prominent in peripheral lymphocytes, thymus, skeletal muscle, and brain. The full CD100 protein is 862 amino acids with homolgy to the semaphorin family of proteins at its extracellular domain followed by a short Ig-like domain. The short cytoplasmic domain contains a putative tyrosine phosphorylation site which may be the site for association with intracellular kinases. Cells that overexpress CD100 induce a rapid aggregation of B cells and increase the CD90L-B cell aggregation. Because semaphorins are axon-guiding molecules for the nervous system, it has been proposed that CD100 may have a guidance role in the immune system.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD100
Applications Western Blot
Classification Monoclonal
Clone 30
Concentration 250μg/mL
Conjugate Unconjugated
Description Semaphorin 4D; Sema 4D
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.