Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD101 (Igsf2) Rat anti-Mouse, Alexa Fluor 647, Clone: 307707, BD

Catalog No. BDB564473 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB564473 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB564473 Supplier BD Biosciences Supplier No. 564473
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The 307707 monoclonal antibody specifically binds to CD101 which is also known as Igsf2 (Immunoglobulin superfamily member 2). CD101 is a type I transmembrane glycoprotein that belongs to the EWI (sharing a conserved glutamine-tryptophan-isoleucine motif) family within the Ig superfamily. CD101 is expressed on monocytes, granulocytes, dendritic cells, Langerhans cells and activated T cells. CD101 reportedly plays a role in the regulation of T cell activation. CD101 was found to be expressed on a subset of CD25+Foxp3+ T regulatory cells that showed higher suppressive properties in vitro and in vivo. Polymorphisms in Cd101 are associated with type I diabetes susceptibility.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD101
Applications Flow Cytometry
Classification Monoclonal
Clone 307707
Conjugate Alexa Fluor 647
Description Igsf2; EWI-101; Gm1016; Gm734
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Rat
Immunogen Mouse CD101 Recombinant Protein
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.