Learn More
CD102 Mouse anti-Human, APC, Clone: CBR-IC2/2, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 564677
Description
The CBR-IC2/2 monoclonal antibody specifically binds to CD102 which is also known as, intercellular adhesion molecule-2 (ICAM-2/ICAM2). CD102 is a type I transmembrane glycoprotein, member of the immunoglobulin supergene family, with an approximate molecular weight of 55-65 kDa. Its extracellular domain consists of two C2-type immunoglobulin-like subunits. The transmembrane region is 26 residues in size and the intracellular region also has 26 residues. CD102 is expressed on vascular endothelial cells, lymphocytes, monocytes, eosinophils, and platelets, but not on resting neutrophils. It is a ligand for the leukocyte integrin CD11a/CD18, or leukocyte function-associated antigen-1 (LFA-1), and there are reports of CD102 binding to leukocyte integrin CD11b/CD18 (Mac-1). Antibody CBR-IC2/2 blocks the binding of CD102 to leukocyte integrin CD11a/CD18. CD102 plays an important role in lymphocyte recirculation and in providing costimulatory signals in the immune response.

Specifications
CD102 | |
Monoclonal | |
APC | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Human ICAM-2 Transfected Cell Line | |
50 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
CBR-IC2/2 | |
ICAM2; ICAM-2; Intercellular adhesion molecule 2 | |
Mouse | |
Affinity Purified | |
RUO | |
Human | |
IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.