Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD103 Mouse anti-Human, APC, Clone: Ber-ACT8, BD

Catalog No. BDB563883 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB563883 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB563883 Supplier BD Biosciences Supplier No. 563883
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The Ber-ACT8 monoclonal antibody specifically binds to the human mucosal lymphocyte antigen 1 (HML-1). This is a 175 kDa type I transmembrane glycoprotein, also known as integrin αE (ITGAE, Integrin alpha-E) and CD103. It is found on >90% of intestinal intraepithelial lymphocytes (iIEL) associated with integrin β7. CD103 is also expressed on lamina propria T lymphocytes in the intestine and on phytohemagglutinin-stimulated peripheral blood lymphocytes. It is rarely expressed on resting peripheral blood lymphocytes. It has been suggested that CD103 may have an accessory function for activation of iIEL. CD103 has been reported as a useful tool in hairy cell leukemia research.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD103
Applications Flow Cytometry
Classification Monoclonal
Clone Ber-ACT8
Conjugate APC
Description ITGAE; Integrin alpha-E; ITAE; HML-1 antigen; Mucosal lymphocyte 1; HUMINAE
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human MAPS16 T Cell Line
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.