Learn More
CD103 Mouse anti-Human, FITC, Clone: BER-ACT8, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 340945
Description
The CD103 antibody, clone Ber-ACT8, is derived using standard cell hybridization procedures using spleen cells from BALB/c mice immunized with the HTLV-1–positive T-cell line, MAPS16.
The CD103 antibody recognizes the αE subunit of integrin αEβ7, an integrin also known as the human mucosal lymphocyte (HML) antigen 1, but not α4, another member of the β7 integrin subfamily. Integrin αEβ7 is a trimeric protein complex of three components, 105 kilodaltons (kDa) (β7), 150 kDa, and 25 kDa. The CD103 antibody recognizes an epitope localized on the 150-kDa chain.

Specifications
CD103 | |
Monoclonal | |
25μg/mL | |
Phosphate buffered saline with gelatin and sodium azide. | |
Human MAPS16 T Cell Line | |
RUO | |
Primary | |
Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed. |
Flow Cytometry | |
BER-ACT8 | |
FITC | |
Mouse | |
50 Tests | |
Immunology | |
Human | |
IgG1 κ |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.