Learn More
CD103 Rat anti-Mouse, FITC, Clone: M290, BD
Rat Monoclonal Antibody
Supplier: BD Biosciences 557494
Description
The M290 antibody specifically binds to CD103, the α chain of αIELβ7 integrin. CD103 has a unique and fairly restricted tissue distribution. It is expressed on almost all intestinal intraepithelial lymphocytes (IEL), on dendritic epidermal T cells (DEC), on subpopulations of peripheral T cells, and on distinct subsets of fetal, neonatal, and adult thymocytes. E-cadherin is the epithelial-cell ligand for αIELβ7 integrin. The ordered expression of αIEL during thymocyte development (which occurs under the influence of the thymic epithelium), the high level of expression of ααIELβ7-mediated adhesion in in vitro assays.

Specifications
CD103 | |
Monoclonal | |
0.5mg/mL | |
Itgae; Integrin alpha-E; αE; alpha-E1; ITAE; Integrin αIEL chain; aM290 | |
Rat | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
M290 | |
FITC | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse Intestinal Epithelial Cells | |
0.1 mg | |
Immunology | |
Mouse | |
IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.