Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD105 Mouse anti-Human, FITC, Clone: 266, BD

Catalog No. BDB561443 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB561443 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB561443 Supplier BD Biosciences Supplier No. 561443
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 266 monoclonal antibody specifically binds to CD105. CD105 presents an integral membrane homodimer protein with subunits of 95 kDa found on vascular endothelial cells and syncytiotrophoblasts of placenta. CD105 is weakly expressed on stromal fibroblasts. It is also expressed on U937 cells, activated macrophages, and mesenchymal stem cells. CD105 is a component of the TGF-β receptor system in human umbilical vein endothelial cells and binds TGF-β1 and TGF-β3 with high affinity but does not bind toTGF-β2. Expression of CD105 is increased on activated endothelium in tissues undergoing angiogenesis, such as in tumors, or in cases of wound healing or dermal inflammation.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD105
Applications Flow Cytometry
Classification Monoclonal
Clone 266
Conjugate FITC
Description Endoglin; ENG; END; HHT1; ORW; ORW1
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human Umbilical Vein Endothelial Cells
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.