Learn More
CD106 Rat anti-Mouse, Alexa Fluor 647, Clone: 429 (MVCAM.A), BD
Catalog No. BDB561612 Shop All BD Biosciences Products
Change view
Click to view available options
50μg; Alexa Fluor 647
Catalog No.
BDB561612 50μg; Alexa Fluor 647
1 options
This item is not returnable. View return policy
Catalog No. BDB561612 Supplier BD Biosciences Supplier No. 561612
Add to Cart
Add to Cart
This item is not returnable. View return policy
Rat Monoclonal Antibody
The 429 monoclonal antibody specifically binds to both the long (∽110 kDa) transmembrane-spanning form and the truncated (∽47 kDa) GPI-linked form of vascular cell adhesion molecule-1 (VCAM-1, CD106). CD106 is constitutively expressed on bone marrow stromal cells, myeloid cells, and splenic dendritic cells. Its expression on endothelial cells is upregulated by inflammatory cytokines and in certain pathologic conditions. CD106 expression has also been detected on apoptotic thymocytes, splenocytes, and lymphoid cell lines. VCAM-1 is a counter-receptor for VLA-4 ( α 4β 1 integrin) and LPAM-1 ( α 4β 7 integrin), and the 429 antibody partially blocks VCAM-1-mediated binding functions. Source of the immunogen was the mouse preadipose cell line PA6.