Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD106 Rat anti-Mouse, Unlabeled, Clone: 429 (MVCAM.A), BD

Catalog No. BDB550547 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB550547 1 mL
BDB553330 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB550547 Supplier BD Biosciences Supplier No. 550547
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The 429 monoclonal antibody specifically binds to both the long (∼110 kDa) transmembrane-spanning form and the truncated (∼47 kDa) GPI-linked form of vascular cell adhesion molecule-1 (VCAM-1, CD106). CD106 is constitutively expressed on bone marrow stromal cells, myeloid cells, and splenic dendritic cells. Its expression on endothelial cells is upregulated by inflammatory cytokines and in certain pathologic conditions. CD106 expression has also been detected on apoptotic thymocytes, splenocytes, and lymphoid cell lines. VCAM-1 is a counter-receptor for VLA-4 ( α; 4 β; 1 integrin) and LPAM-1 ( α; 4 β; 7 integrin), and the 429 antibody partially blocks VCAM-1-mediated binding functions. Source of the immunogen was the mouse preadipose cell line PA6.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD106
Applications Flow Cytometry
Classification Monoclonal
Clone 429 (MVCAM.A)
Concentration 31.25μg/mL
Conjugate Unconjugated
Description Vcam-1; Vascular cell adhesion molecule 1; Vascular cell adhesion protein 1
Formulation Aqueous buffered solution containing BSA, goat serum, and ≤0.09% sodium azide.
Host Species Rat
Immunogen Mouse preadipose cell line PA6
Purification Method Affinity Purified
Quantity 1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.