Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD106 Mouse anti-Rat, PE, Clone: MR106, BD

Catalog No. BDB559229 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB559229 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB559229 Supplier BD Biosciences Supplier No. 559229
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The MR106 antibody reacts with CD106, the ∽110 kDa transmembrane vascular cell adhesion molecule-1 (VCAM-1). Expression of CD106 on endothelial cells is induced by inflammatory cytokines and in certain pathologic conditions. In the rat bone marrow, CD106 has been reported to be expressed on dendritic cells and subsets of the myeloid lineage, but not detectable on peripheral leukocytes. CD106 has been reported to be a counter-receptor for VLA-4 ( α 4β 1 integrin) and α 4β 7 integrin, and it plays an important role in leukocyte extravasation, particularly at sites of inflammation.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD106
Applications Flow Cytometry
Classification Monoclonal
Clone MR106
Concentration 0.2mg/mL
Conjugate PE
Description VCAM-1
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Rat VCAM-1-transfected L5178Y Cells
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Rat
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.