Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD107a Rat anti-Mouse, Brilliant Violet 711, Clone: 1D4B, BD

Catalog No. BDB564348 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB564348 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB564348 Supplier BD Biosciences Supplier No. 564348
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The 1D4B antibody recognizes CD107a (Lysosome-Associated Membrane Protein 1, LAMP-1), one of the two major glycoproteins in lysosome membranes which are useful markers to distinguish lysosomes from other organelles. Mouse CD107a consists of a 40-kDa core protein which is heavily glycosylated to form heterogeneous mature glycoprotein of 110-140 kDa. It is principally expressed in epithelial cells and macrophages in a variety of organs in normal and Beige mutant mice. Several lines of evidence suggest that cell-surface mouse and human LAMP-1 glycoproteins, the expression of which may correlate with malignant transformation, participate in intercellular adhesion and adhesion to extracellular matrix.

The antibody was conjugated to BD Horizon™ BV711 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 711-nm. BD Horizon BV711 can be excited by the violet laser and detected in a filter used to detect Cy™5.5 / Alexa Fluor™ 700-like dyes (eg, 712/20-nm filter). Due to the excitation and emission characteristics of the acceptor dye, there may be moderate spillover into the Alexa Fluor™ 700 and PerCP-Cy5.5 detectors. However, the spillover can be corrected through compensation as with any other dye combination.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD107a
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 1D4B
Concentration 0.2mg/mL
Conjugate Brilliant Violet 711
Description LAMP-1; LGP-120; LGP-A; P2B
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Rat
Immunogen Plasma membrane fraction of mouse embryo NIH 3T3 cell line
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.