Learn More
CD107a Mouse anti-Human, PE-Cy5, Clone: H4A3, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 555802
Description
Reacts with the heavily glycosylated 110kDa lysosomal-associated membrane protein, LAMP-1. LAMP-1 is a widely expressed intracellular antigen. It is also expressed on the surface of activated platelets, PHA-activated lymphocytes and some tumor cell lines, including U937 and KG1a. LAMP-1 has been shown to be a ligand for E-selectin-mediated cell adhesion. LAMP-1 and LAMP-2 (CD107b) are carriers for poly-N-acetyllactosamines, and are able to display sialyl Le[x] termini. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Mouse
Clone: H4A3
Isotype: IgG1 κ
Species Reactivity: Human
Intracellular Staining

Specifications
CD107a | |
Monoclonal | |
PE-Cyanine5 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Human Adult Adherent Peripheral Blood Cells | |
20 μL/Test | |
Immunology | |
Human | |
IgG1 κ |
Flow Cytometry | |
H4A3 | |
LAMP1; LAMP-1; LAMPA; LGP120 | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
100 Tests |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.