Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD107b Mouse anti-Human, FITC, Clone: H4B4, BD

Catalog No. BDB560946 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB560946 25 Tests
BDB555804 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB560946 Supplier BD Biosciences Supplier No. 560946
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reacts with the heavily glycosylated 120kDa lysosmal-associated protein, LAMP-2. LAMP-2 is a widely expressed intracellular antigen. It is also expressed on the surface of activated platelets, PHA-activated lymphocytes and some tumor cell lines, including U937 and KG1a. LAMP-2 has been shown to be a ligand for E-selectin-mediated cell adhesion. LAMP-1 (CD107a) and LAMP-2 are carriers for poly-N-acetyllactosamines, and are able to display sialyl Le[x] termini.

Host Species: Mouse
Clone: H4B4
Isotype: IgG1 κ
Species Reactivity: Human

Intracellular Staining

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD107b
Applications Flow Cytometry
Classification Monoclonal
Clone H4B4
Conjugate FITC
Description LAMP-2
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human Adult Adherent Peripheral Blood Cells
Purification Method Affinity Purified
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.