Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD107b Rat anti-Mouse, PerCP-Cy5.5, Clone: M3/84, BD

Catalog No. BDB564844 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB564844 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB564844 Supplier BD Biosciences Supplier No. 564844
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The M3/84 monoclonal antibody specifically binds to CD107b which is also known as Mac-3, Lysosome-associated membrane protein 2 (LAMP-2/Lamp2/Lamp II), and Lysosomal membrane glycoprotein type B (LGP-B). CD107b is a single-pass type I transmembrane glycoprotein that constitutes a major integral membrane protein of lysosomes and may play a role in lysosomal function. CD107b is also expressed on the surface of mouse mononuclear phagocytes. Surface expression of the 92-110-kDa glycoprotein antigen increases during differentiation of monocytes to activated macrophages and may play a role in adhesion. The M3/84 mAb can detect CD107b antigen on tissue macrophages, thioglycollate-elicited peritoneal macrophages, and some myeloid cell lines, but not on lymphocytes or monocytes. In the bone marrow, very few cells display CD107b antigen on the surface, but a large proportion express cytoplasmic CD107b. The M3/84 antibody has also been reported to stain dendritic cells and endothelium in sections of thymus (both medulla and cortex), lymph nodes, spleen (white pulp), and gut-associated lymphoid tissue.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD107b
Applications Flow Cytometry
Classification Monoclonal
Clone M3/84
Concentration 0.2mg/mL
Conjugate PerCP-Cyanine5.5
Description Mac-3; Lamp2; LAMP-2; Lysosome-associated membrane glycoprotein 2; LGP-B
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Mouse C57Bl/6 peritoneal exudate cells
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.