Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD107b Rat anti-Mouse, PE, Clone: ABL-93, BD

Catalog No. BDB564246 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB564246 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB564246 Supplier BD Biosciences Supplier No. 564246
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The ABL-93 monoclonal antibody specifically recognizes CD107b which is also known as Lysosome-associated membrane protein 2 (LAMP2/LAMP-2), Lysosomal membrane glycoprotein type B (LGP-B), and Mac-3. CD107b is a type I lysosomal transmembrane glycoprotein that serves as a useful marker to distinguish lysosomes from other cellular organelles. Mouse CD107b consists of a ∽40-kDa core protein which is heavily glycosylated to form heterogeneous mature glycoproteins of 110-120 kDa. It is principally expressed in epithelial cells and macrophages in a variety of organs in normal and Beige mutant mice.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD107b
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone ABL-93
Concentration 0.2mg/mL
Conjugate PE
Description Lamp2; LAMP-2; LAMP II; Lysosomal-associated membrane protein 2; LGP-B
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Glycoproteins purified from BALB/c mouse embryo 3T3 cell line
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a λ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.