Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD108 Mouse anti-Human, PE, Clone: KS-2, BD

Catalog No. BDB552830 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB552830 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB552830 Supplier BD Biosciences Supplier No. 552830
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reacts with the John-Milton-Hagen (JMH) blood group antigen. CD108 is a 76-80 kDa glycosylphosphatidylinositol (GPI)-linked protein. It is expressed on erythrocytes, at low levels on lymphocytes, but higher levels are detected on lymphoblasts and lymphoblastic cell lines. An adhesion function for CD108 has been suggested but this has not been demonstrated. Autoantibodies to this molecule are not uncommon and often associated with the loss of JMH antigen expression on erythrocytes.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD108
Applications Flow Cytometry
Classification Monoclonal
Clone KS-2
Conjugate PE
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.