Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD112 Mouse anti-Human, PE, Clone: R2.525, BD

Catalog No. BDB551057 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB551057 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB551057 Supplier BD Biosciences Supplier No. 551057
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reacts with the poliovirus receptor 2 (PRR2) protein. PRR2 is a cell surface molecule homologous to the poliovirus receptor (PVR/CD155). It is a transmembrane glycoprotein belonging to the immunoglobulin superfamily. The PRR2 gene encodes for two mRNA isoforms, a short form of 3.0 kb (hPRR2 α ) and a long form of 4.4 kb (hPRR2 δ ). Both are widely expressed in human tissues, including hematopoietic cells. PRR2 is also expressed on endothelial cells, at the intercellular junctions of adjacent cells. It has been suggested that PRR2 may play a role in the regulation of hematopoietic/endothelial cell interaction.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD112
Applications Flow Cytometry
Classification Monoclonal
Clone R2.525
Conjugate PE
Description PRR2
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.