Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD114 Mouse anti-Human, Unlabeled, Clone: LMM741, BD

Catalog No. BDB554536 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB554536 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB554536 Supplier BD Biosciences Supplier No. 554536

Mouse Monoclonal Antibody

The LMM741 antibody reacts with the 150 kD human granulocyte-colony stimulating factor receptor (G-CSFR) also known as CD114. Cell surface G-CSFR is expressed by neutrophils, myeloid leukemias, endothelium, platelets, and placenta. The immunogen used to generate this hybridoma was cells transfected with an expression vector containing a full-length cDNA encoding the G-CSF-receptor.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD114
Applications Flow Cytometry
Classification Monoclonal
Clone LMM741
Concentration 0.5mg/mL
Conjugate Unconjugated
Description G-CSF Receptor
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen G-CSFR cDNA transfected cells.
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.