Learn More
BioLegend™ anti-CD117, Clone: 2B8
Description
Host Species: Rat
Clone: 2B8
Isotype: IgG2b, κ
Cross Reactivity: Mouse
Immunogen: Mouse bone marrow mast cells
Flow Cytometry
Specifications
Specifications
| Antigen | CD117 |
| Classification | Monoclonal |
| Clone | 2B8 |
| Concentration | 0.2mg/mL |
| Conjugate | APC |
| Host Species | Rat |
| Immunogen | Mouse bone marrow mast cells |
| Purification Method | Unpurified |
| Quantity | 100 μg |
| Primary or Secondary | Primary |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.