Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD117 Rat anti-Mouse, Clone: ACK2, MilliporeSigma™

Catalog No. CBL1360MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CBL1360MI 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CBL1360MI Supplier MilliporeSigma Supplier No. CBL1360
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Detect CD117 using this Anti-CD117 Antibody, clone ACK2 validated for use in flow cytometry, immunohistochemistry, immunoprecipitation, FUNC and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD117
Applications Flow Cytometry, Functional Assay, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone ACK2
Conjugate Unconjugated
Formulation The monoclonal is presented in phosphate buffered saline containing 10mM sodium azide.
Gene Accession No. NM_000222.2; NM_001093772.1
Gene Symbols KIT, CD117, SCFR, C-Kit, PBT, c-kit, EC 2.7.10.1
Host Species Rat
Purification Method Protein A purfied
Quantity 500 μg
Primary or Secondary Primary
Target Species Mouse
Content And Storage 4°C for up to 1 month, −20°C in undiluted aliquots for long term
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.