Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD117 Mouse anti-Human, PE, Clone: 104D2, BD

Catalog No. BDB340529 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB340529 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB340529 Supplier BD Biosciences Supplier No. 340529
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD117, clone 104D2, is derived from fusion of Sp2/0 myeloma cells with splenocytes from BALB/c mice immunized with the megakaryoctic cell line MOLM-1.

CD117 binds to the receptor for stem cell factor (SCF). It selectively recognizes NIH-3T3 cells transfected with human c-kit, the gene that codes for SCF-R. CD117 antibody does not block the epitope that binds SCF.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD117
Applications Flow Cytometry
Classification Monoclonal
Clone 104D2
Concentration 10μg/mL
Conjugate PE
Formulation Phosphate buffered saline with gelatin and 0.1% sodium azide.
Host Species Mouse
Immunogen Megakaryoctic cell line MOLM-1
Quantity 50 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.