Learn More
CD117 Mouse anti-Human, PE-Cy7, Clone: 104D2, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 339195
Description
CD117, clone 104D2, is derived from fusion of Sp2/0 myeloma cells with splenocytes from BALB/c mice immunized with the megakaryoctic cell line MOLM-1.
CD117 binds to the receptor for stem cell factor (SCF). It selectively recognizes NIH-3T3 cells transfected with human c-kit, the gene that codes for SCF-R. CD117 antibody does not block the epitope that binds SCF.

Specifications
CD117 | |
Monoclonal | |
12.5μg/mL | |
Phosphate buffered saline with gelatin and 0.1% sodium azide. | |
Megakaryoctic cell line MOLM-1 | |
RUO | |
Primary | |
Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed. |
Flow Cytometry | |
104D2 | |
PE-Cyanine7 | |
Mouse | |
100 Tests | |
Immunology | |
Human | |
IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.