Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD119 Hamster anti-Mouse, Biotin, Clone: 2E2, BD

Catalog No. BDB550482 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB550482 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB550482 Supplier BD Biosciences Supplier No. 550482
Add to Cart
Add to Cart

Hamster Monoclonal Antibody

The 2E2 antibody recognizes the extracellular region of the 90 kDa alpha chain subunit of the mouse interferon- γreceptor (IFN- γR α ; aka , CD119). The functionally active-form of the mouse IFN- γreceptor consists of two (or more) subunits, with IFN- γγR α and IFN- γR β chains required for the transduction of biologic responses. IFN- γR α is expressed by a variety of cell lines and normal mouse cells (except mature erythrocytes) including T cells, B cells, NK cells, monocytes, neutrophils, fibroblasts, epithelial and endothelial cells. The 2E2 antibody is a non-neutralizing antibody; it does not block the binding of IFN- γto its receptor. The immunogen used to generate this hybridoma was a purified preparation of soluble recombinant mouse IFN- γR α chain protein.

This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD119
Applications Flow Cytometry
Classification Monoclonal
Clone 2.00E+02
Concentration 0.5mg/mL
Conjugate Biotin
Description Ifngr1; IFN-γ Receptor α; IFN-gamma R alpha; Ifngr; INF-g Receptor; Cd119
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Hamster
Immunogen Purified preparation of soluble recombinant mouse IFN-γRα chain protein
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.