Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD119 Hamster anti-Mouse, Unlabeled, Clone: 2E2., BD

Catalog No. BDB559911 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB559911 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB559911 Supplier BD Biosciences Supplier No. 559911

Hamster Monoclonal Antibody

The 2E2 antibody recognizes the extracellular region of the 90 kDa alpha chain subunit of the mouse interferon- γreceptor (IFN- γR α ; aka , CD119). The functionally active-form of the mouse IFN- γreceptor consists of two (or more) subunits, with IFN- γγR α and IFN- γR β chains required for the transduction of biologic responses. IFN- γR α is expressed by a variety of cell lines and normal mouse cells (except mature erythrocytes) including T cells, B cells, NK cells, monocytes, neutrophils, fibroblasts, epithelial and endothelial cells. The 2E2 antibody is a non-neutralizing antibody; it does not block the binding of IFN- γto its receptor. The immunogen used to generate this hybridoma was a purified preparation of soluble recombinant mouse IFN- γR α chain protein.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD119
Applications Flow Cytometry
Classification Monoclonal
Clone 2E2.
Concentration 0.5mg/mL
Conjugate Unconjugated
Description IFN-γ Receptor α chain, IFN gamma Receptor alpha chain, IFN y R alpha chai
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Hamster
Immunogen Purified preparation of soluble recombinant mouse IFN-γRα chain protein
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.