Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD119 Mouse anti-Human, PE, Clone: GIR-208, BD

Catalog No. BDB558934 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558934 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558934 Supplier BD Biosciences Supplier No. 558934
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The GIR-208 antibody recognizes the extracellular region of CD119 which is also known as the alpha chain subunit (80-95 kDa glycoprotein) of the human interferon- γreceptor (IFN- γR α ). The functionally active-form of the human IFN- γreceptor consists of two (or more) subunits, with IFN- γR α responsible for IFN- γβ chains required for the transduction of biologic responses. The IFN- γreceptor α chain (CD119) is expressed on the surface of most human cells (except mature erythrocytes) including monocytes, macrophages, T cells, B cells, NK cells, neutrophils, fibroblasts, epithelial cells, and endothelium. Binding of 125I-labeled GIR-208 antibody to IFN- γR α + cells is reported to be specifically inhibited in the presence of excess IFN- γ. GIR-208 does not cross react with IFN- γas tested by ELISA. The ability of this antibody to bind to IFN- γreceptors of species other than human has not been determined. The immunogen used to generate this hybridoma was human IFN- γR α purified from human placenta. The GIR- 208 has been reported to block the binding of 125I-human IFN-γto IFN-γRα+ cells as well as purified, soluble human IFN-γRα. GIR-208 is a neutralizing antibody that has been shown to neutralize the anti-viral activity of IFN-γon WISH cells in a dose-dependent fashion.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD119
Applications Flow Cytometry
Classification Monoclonal
Clone GIR-208
Concentration 0.2mg/mL
Conjugate PE
Description IFN-γ Receptor α chain, IFNγRα
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human IFN-γRα
Purification Method Affinity Purified
Quantity 0.2 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.