Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD11a Mouse anti-Rat, Unlabeled, Clone: WT.1, BD

Catalog No. BDB559979 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB559979 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB559979 Supplier BD Biosciences Supplier No. 559979

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

The WT.5 antibody reacts with the asubunit of LFA-1 (αLβ2 integrin, CD11a/CD18), a heterodimeric surface glycoprotein which is found on the majority of leukocytes, but not on peritoneal macrophages or peritoneal mast cells. LFA-1 mediates a variety of heterotypic and homotypic intercellular adhesions through interaction with ICAM-1 (CD54) and ICAM-2 (CD102). WT.1 mAb recognizes both the activated and unactivated forms of LFA-1. It inhibits the binding of LFA-1 to ICAM-1 in several in vitro assays, including binding of Concanavalin A-stimulated lymphocytes (Con A blasts) to purified ICAM-1 and Mg2+-dependent aggregation of concanavalin A-stimulated blasts. It has also been reported to inhibit leukocyte infiltration in several in vivo models of inflammation.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD11a
Applications Flow Cytometry
Classification Monoclonal
Clone WT.1
Concentration 0.5mg/mL
Conjugate Unconjugated
Description Integrin αL chain, LFA-1 α chain
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen PHA-stimulated rat splenocytes and rat thymic lymphoma FTL-43
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Rat
Content And Storage Store undiluted at 4°C
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.