Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD11b Rat anti-Mouse, APC-Cy7, Clone: M1/70, BD

Catalog No. BDB561039 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB561039 25 μg
BDB557657 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB561039 Supplier BD Biosciences Supplier No. 561039
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The M1/70 antibody reacts with the 170kDa α[M] chain of Mac-1 (CD11b/CD18, α[M]β[2] integrin), also known as complement receptor 3 (CR3), which mediates adhesion to C3bi and ICAM-1 (CD54). Mac-1 is expressed at varying levels on granulocytes, macrophages, myeloid-derived dendritic cells, natural killer cells, microglia, and B-1 cells. Mac-1 expression is rapidly up-regulated on neutrophils after activation, in the same time period that CD62L (L-selectin) is shed from the cell surface. M1/70 antibody reportedly blocks cell adherence and C3bi binding, but it does not block cell-mediated lysis. Cross-reaction of mAb M1/70 with CD11b on human monocytes, polymorphonuclear leukocytes, and NK cells has been reported. This antibody is routinely tested by flow cytometric analysis (≤0.25 μg/million cells).

Host Species: Rat
Clone: M1/70
Isotype: IgG2b, κ
Species Reactivity: Mouse
Immunogen: Mouse splenic Cells

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD11b
Applications Flow Cytometry
Classification Monoclonal
Clone M1/70
Concentration 0.2mg/mL
Conjugate APC-Cyanine7
Description Itgam; Integrin alpha-M; Ly-40; Mac-1a; Mac-1 alpha; CR3A; CR-3 alpha chain
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Mouse Splenic Cells
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.