Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BioLegend™ CD11b, Rat anti-Mouse, Biotin, Clone: M1/70

Catalog No. NC0200884
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NC0200884 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NC0200884 Supplier BioLegend™ Supplier No. 101204
Add to Cart
Add to Cart

Specifications

Antigen CD11b
Classification Monoclonal
Clone M1/70
Concentration 0.5mg/mL
Conjugate Biotin
Host Species Rat
Immunogen C57BL/10 splenocytes
Purification Method Unpurified
Quantity 500 μg
Primary or Secondary Primary
Target Species Mouse
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.