Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD11b (CR3) Mouse anti-Human, PE, Clone: D12, BD

Catalog No. BDB347557 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB347557 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB347557 Supplier BD Biosciences Supplier No. 347557
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD11b, clone D12, is derived from hybridization of mouse NS-1 myeloma cells with spleen cells from BALB/c mice immunized with peripheral blood T lymphocytes.

CD11b recognizes a human leucocyte antigen that is the C3bi complement receptor (CR

3). CD11b is specific for the Mr 165-kilodalton (kd) α subunit of the CD11b/CD18 antigen heterodimer. The Mr 95-kd CD18 antigen, the β chain of leucocyte function–associated antigen-1 (LFA-1β), is common to the CD11a/CD18 (LFA-1) and CD11c/CD18 antigen heterodimers.

3). CD11b is specific for the Mr 165-kilodalton (kd) α subunit of the CD11b/CD18 antigen heterodimer. The Mr 95-kd CD18 antigen, the β chain of leucocyte function–associated antigen-1 (LFA-1β), is common to the CD11a/CD18 (LFA-1) and CD11c/CD18 antigen heterodimers.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD11b
Applications Flow Cytometry
Classification Monoclonal
Clone D12
Concentration 50μg/mL
Conjugate PE
Formulation Phosphate buffered saline with gelatin and 0.1% sodium azide.
Host Species Mouse
Immunogen Peripheral blood T lymphocytes
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.