Learn More
CD11b/Mac-1 Mouse anti-Human, PE, Clone: ICRF44 (also known as 44), BD
Shop All BD Biosciences ProductsDescription
The ICRF44 monoclonal antibody specifically binds to CD11b, the 165-kDa adhesion glycoprotein that associates with the 95-kDa integrin β2 (CD18) to form the CD11b/CD18 complex, also known as Mac-1 or CR3. CD11b is expressed on activated lymphocytes, monocytes, granulocytes, and a subset of NK cells. CD11b functions in cell-cell and cell-substrate interactions and is a receptor for iC3b, CD54 (ICAM-1), CD102 (ICAM-2) and CD50 (ICAM-3). This antibody significantly inhibits polymorphonuclear leukocyte aggregation in response to fMLP.
Specifications
Specifications
| Antigen | CD11b |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | ICRF44 (also known as 44) |
| Conjugate | PE |
| Description | Mac-1α, integrin αM subunit, CR3 α chain |
| Formulation | Aqueous buffered solution containing BSA and ≤0.09% sodium azide. |
| Host Species | Mouse |
| Immunogen | Human monocytes |
| Purification Method | Affinity Purified |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.