Learn More
CD11c Mouse anti-Human, APC, Clone: B-LY6, BD
Mouse Monoclonal Antibody
$162.70 - $404.50
Specifications
| Antigen | CD11c |
|---|---|
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | APC |
| Host Species | Mouse |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
BDB560895
|
BD Biosciences
560895 |
25 Tests |
Each for $162.70
|
|
|||||
|
BDB559877
|
BD Biosciences
559877 |
100 Tests |
Each for $404.50
|
|
|||||
Description
Reacts with the 150kDa adhesion glycoprotein (p150, integrin α chain) expressed on NK cells, a subset of B and T cells, monocytes, granulocytes and macrophages. It associates with CD18 to form the CD11c/CD18 complex, which binds fibrinogen and has been reported to be a receptor for iC3b and ICAM-1. Reports indicate that CD11c/CD18 may play a role as an adhesion molecule which binds to receptors on stimulated epithelium. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Flow Cytometry
Specifications
| CD11c | |
| Monoclonal | |
| Mouse | |
| Human | |
| Spleen Cells from Human with Hairy Cell Leukemia | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| APC | |
| RUO | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| IgG1 κ | |
| Affinity Purified | |
| ITGAX; AlphaX integrin chain; Axb2; Integrin alpha-X; CR4; Leu M5; SLEB6 |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.