Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CD11c APC-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 1 publication
$210.00 - $432.50
Specifications
Antigen | CD11c |
---|---|
Clone | ICRF 3.9 |
Dilution | Flow Cytometry 10 uL/10^6 cells |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FAB1777A025
![]() |
R&D Systems
FAB1777A025 |
25 Tests |
Each for $210.00
|
|
|||||
FAB1777A100
![]() |
R&D Systems
FAB1777A100 |
100 Tests |
Each for $432.50
|
|
|||||
Description
CD11c Monoclonal specifically detects CD11c in Human samples. It is validated for Flow Cytometry.Specifications
CD11c | |
Flow Cytometry 10 uL/10^6 cells | |
Monoclonal | |
Purified | |
RUO | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
3687 | |
Rheumatoid synovial fluid cells and human monocyte-derived fibronectin | |
Primary | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
ICRF 3.9 | |
Flow Cytometry | |
APC | |
Mouse | |
Human | |
95, 95 alpha chain, 95, alpha subunit, CD11 antigen-like family member C, CD11c, CD11c antigen, CD11Cleu M5, alpha subunit, Integrin alpha X, integrin alpha-X, integrin, alpha X (antigen CD11C (p150), alpha polypeptide), integrin, alpha X (complement component 3 receptor 4 subunit), ITGAX, Leu M5, Leukocyte adhesion glycoprotein p150, Leukocyte adhesion receptor p150, leukocyte surface antigen p150, myeloid membrane antigen, alpha subunit, p150 95 integrin alpha chain, p150,95 alpha, SLEB6 | |
ITGAX | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Detects human CD11c. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title