Learn More
CD11c Hamster anti-Mouse, Biotin, Clone: HL3, BD
Hamster Monoclonal Antibody
Supplier: BD Biosciences 553800
Description
The HL3 monoclonal antibody specifically binds to the integrin αx chain of gp150, 95 (CD11c/CD18). CD11c is expressed on dendritic cells, CD4- CD8+ intestinal intraepithelial lymphocytes (IEL) and some NK cells. It is upregulated on IEL and lymph-node T cells following in vivo activation. Cells of the monocyte/macrophage lineage have been reported to express low levels of CD11c. CD11c plays a role in binding of iC3b.

Specifications
CD11c | |
Monoclonal | |
0.5mg/mL | |
Cd11c; Itgax; Integrin alpha-X; Integrin αX; Cr4; Complement receptor 4 | |
Hamster | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
HL3 | |
Biotin | |
Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide. | |
C57BL/6 Mouse Intestinal Intraepithelial Lymphocytes | |
0.5 mg | |
Immunology | |
Mouse | |
IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.