Learn More
CD11c Mouse anti-Human, Brilliant Violet 605, Clone: B-ly6, BD
Mouse Monoclonal Antibody
$233.00 - $444.00
Specifications
Antigen | CD11c |
---|---|
Clone | B-ly6 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | Brilliant Violet 605 |
Description
The B-ly6 monoclonal antibody specifically binds to the 150 kDa adhesion glycoprotein CD11c (p150, integrin α chain). CD11c is expressed on dendritic cells, monocytes, macrophages, granulocytes, NK cells and subsets of B and T cells. It associates with CD18 to form the CD11c/CD18 complex that binds fibrinogen and has been reported to be a receptor for iC3b and ICAM-1. Reports indicate that CD11c/CD18 plays a role as an adhesion molecule that mediates cellular binding to ligands expressed on stimulated epithelium and endothelium.
This antibody is conjugated to BD Horizon BV605 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407-nm and Em Max of 602-nm, BD Horizon BV605 can be excited by a violet laser and detected with a standard 610/20-nm filter set. BD Horizon BV605 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an Em max at 605-nm. Due to the excitation of the acceptor dye by the green (532 nm) and yellow-green (561 nm) lasers, there will be significant spillover into the PE and BD Horizon PE-CF594 detectors off the green or yellow-green lasers. BD Horizon BV605 conjugates are very bright, often exhibiting brightness equivalent to PE conjugates and can be used as a third color off of the violet laser.
Specifications
CD11c | |
Flow Cytometry | |
Brilliant Violet 605 | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
IgG1 κ | |
Affinity Purified | |
ITGAX; AlphaX integrin chain; Axb2; Integrin alpha-X; CR4; Leu M5; SLEB6 |
B-ly6 | |
Monoclonal | |
Mouse | |
Human | |
Spleen Cells from Human with Hairy Cell Leukemia | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.