Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-CD11c; Clone: B-ly6; BD
$232.00 - $472.00
Specifications
Antigen | CD11c |
---|---|
Clone | B-ly6 |
Classification | Monoclonal |
Conjugate | Brilliant Ultraviolet 395 |
Host Species | Mouse |
Description
Host Species: Mouse
Clone: B-ly6
Isotype: IgG1, κ
Species Reactivity: Hu
Immunogen: Spleen Cells from Human with Hairy Cell Leukemia
Flow cytometry
Specifications
CD11c | |
Monoclonal | |
Mouse | |
Human | |
Primary |
B-ly6 | |
Brilliant Ultraviolet 395 | |
RUO | |
Spleen Cells from Human with Hairy Cell Leukemia | |
Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title