Learn More
CD11c Mouse anti-Human, Unlabeled, Clone: B-LY6, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 550375
Description
The B-ly6 monoclonal antibody specifically binds to the 150 kDa adhesion glycoprotein CD11c (p150, integrin α chain). CD11c is expressed on dendritic cells, monocytes, macrophages, granulocytes, NK cells and subsets of B and T cells. It associates with CD18 to form the CD11c/CD18 complex that binds fibrinogen and has been reported to be a receptor for iC3b and ICAM-1. Reports indicate that CD11c/CD18 plays a role as an adhesion molecule that mediates cellular binding to ligands expressed on stimulated epithelium and endothelium.

Specifications
CD11c | |
Monoclonal | |
31.25μg/mL | |
ITGAX; AlphaX integrin chain; Axb2; Integrin alpha-X; CR4; Leu M5; SLEB6 | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C |
Flow Cytometry | |
B-LY6 | |
Unconjugated | |
Aqueous buffered solution containing BSA, goat serum, and ≤0.09% sodium azide. | |
Spleen Cells from Human with Hairy Cell Leukemia | |
1 mL | |
Immunology | |
Human | |
IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.