Learn More
BioLegend™ anti-CD11c, Clone: N418
Description
Host Species: Hamster
Clone: N418
Isotype: IgG
Cross Reactivity: Mouse
Flow Cytometry
Specifications
Specifications
| Antigen | CD11c |
| Classification | Monoclonal |
| Clone | N418 |
| Concentration | 0.2mg/mL |
| Conjugate | PE |
| Host Species | Hamster |
| Purification Method | Unpurified |
| Quantity | 200 μg |
| Primary or Secondary | Primary |
| Target Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.