Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD11c (ITGAX, CD11C Integrin αX chain) Mouse anti-Human, APC, Clone: S-HCL-3, BD

Catalog No. BDB340544 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB340544 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB340544 Supplier BD Biosciences Supplier No. 340544
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD11c, clone S-HCL-3, is derived from hybridization of mouse NS-1 myeloma cells with spleen cells from CD-1 mice immunized with whole cells and membrane preparations of hairy-cell leukemia.

CD11c recognizes a human monocyte/macrophage antigen. CD11c is specific for the Mr 150-kDa α-subunit of the CD11c/CD18 antigen heterodimer, also referred to as complement receptor type 4 (CR4). The Mr 95-kDa CD18 antigen, the β chain of leucocyte function-associated antigen-1 (LFA-1), is common to the CD11a/CD18 (LFA-1) and CD11b/CD18 antigen heterodimers.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD11c
Applications Flow Cytometry
Classification Monoclonal
Clone S-HCL-3
Conjugate APC
Formulation Phosphate buffered saline with gelatin and 0.1% sodium azide.
Host Species Mouse
Immunogen Human Hairy-Cell Leukemia Cells
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed.
Isotype IgG2b κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.