Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD11c Hamster anti-Mouse, PE-Cy7, Clone: HL3, BD

Catalog No. BDB561022 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB561022 25 μg
BDB558079 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB561022 Supplier BD Biosciences Supplier No. 561022
Add to Cart
Add to Cart

Hamster Monoclonal Antibody

The HL3 antibody reacts with the integrin αx chain of gp150, 95 (CD11c/CD18) which is expressed on dendritic cells and CD4- CD8+ intestinal intraepithelial lymphocytes (IEL) and is upregulated on IEL and lymph-node T cells following in vivo activation. CD11c is also found on human NK cells. Although its expression on mouse NK cells is not published, we have detected CD11c on mouse splenic NK cells. Cells of the monocyte/macrophage lineage have been reported to express low levels of CD11c. CD11c plays a role in binding of iC3b.

Host Species: Hamster
Clone: HL3
Isotype: IgG1, <special id='310'/>2
Species Reactivity: Mouse
Immunogen: C57BL/6 Mouse Intestinal Intraepithelial Lymphocytes

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD11c
Applications Flow Cytometry
Classification Monoclonal
Clone HL3
Concentration 0.2mg/mL
Conjugate PE-Cyanine7
Description Cd11c; Itgax; Integrin alpha-X; Integrin αX; Cr4; Complement receptor 4
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Hamster
Immunogen C57BL/6 Mouse Intestinal Intraepithelial Lymphocytes
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 λ2
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.