Learn More
CD120a Mouse anti-Human, Unlabeled, Clone: MABTNFR1-A1, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 552535
Description
The MABTNFR1-A1 antibody reacts with the extracellular domain of the 55 kDa receptor for the human cytokines: TNF (aka tumor necrosis factor-alpha , TNF- α ) and lymphotoxin-alpha (LT- α 3, aka, lymphotoxin or TNF- β ). This receptor is referred to as the p55 or Type I Tumor Necrosis Factor Receptor (TNFRI) or CD120a. TNFRI are expressed by a variety of cell lines, tumor cells, and normal cell types including T cells, monocytes, macrophages, neutrophils, endothelial cells, hepatocytes, chondrocytes, and fibroblasts. Naive B cells express very low or undetectable levels of TNFRI whereas mature erythrocytes and platelets are uniformly negative for TNFRI expression. For the generation of the MABTNFRI-B1 hybridoma, BALB/c mice were immunized with purified, full-length human TNFRI protein. MABTNFRI-A1 specifically binds to natural and recombinant truncated forms of TNFRI.

Specifications
CD120a | |
Monoclonal | |
0.5mg/mL | |
TNF Receptor Type I | |
Mouse | |
0.5 mg | |
Immunology | |
Human | |
IgG1 κ |
ELISA | |
MABTNFR1-A1 | |
Unconjugated | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.