Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD120a Mouse anti-Human, Unlabeled, Clone: MABTNFR1-B1, BD

Catalog No. BDB550514 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB550514 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB550514 Supplier BD Biosciences Supplier No. 550514
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The TNFR1-B1 antibody reacts with the extracellular domain of CD120A which is also known as the 55 kDa receptor for the human cytokines, tumor necrosis factor (TNF) and lymphotoxin-alpha (LT- α ). This receptor is referred to as the p55 or Type I Tumor Necrosis Factor Receptor (TNFRI). TNFRI are expressed by a variety of cell lines, tumor cells, and normal cell types including T cells, monocytes, macrophages, neutrophils, endothelial cells, hepatocytes, chondrocytes, and fibroblasts. Naive B cells express very low or undetectable levels of TNFRI whereas mature erythrocytes and platelets are uniformly negative for TNFRI expression. For the generation of the MABTNFR1-B1 hybridoma, BALB/c mice were immunized with purified, full-length human TNFRI protein that was expressed by insect cells that were infected with a recombinant human TNFRI-baculovirus expression vector. MABTNFR1-B1 specifically binds to natural and recombinant truncated forms of TNFRI.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD120a
Applications Flow Cytometry
Classification Monoclonal
Clone MABTNFR1-B1
Concentration 0.5mg/mL
Conjugate Unconjugated
Description TNF Receptor Type I
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.