Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD120b Armenian Hamster anti-Mouse, Brilliant Violet 421, Clone: TR75-89, BD

Catalog No. BDB564088 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB564088 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB564088 Supplier BD Biosciences Supplier No. 564088
Add to Cart
Add to Cart

Armenian Hamster Monoclonal Antibody

The TR75-89 monoclonal antibody specifically binds to the extracellular region of CD120b, the 75 kDa receptor for the mouse cytokines, tumor necrosis factor (TNF, aka TNF- α ) and lymphotoxin-alpha (LT- α , aka lymphotoxin, TNF- β ). This receptor, referred to as the p75 or Type II Tumor Necrosis Factor Receptor (TNFRII), or TNFRSF1B, is expressed by a variety of cell lines and normal cell types including T cells, monocytes, macrophages, and neutrophils. Resting B cells express low or undetectable levels of TNFRII whereas mature erythrocytes are uniformly negative for TNFRII expression. In addition, the TR75-89 antibody can bind to a soluble, truncated form of the mouse Type II TNFR that is shed by cells in response to certain stimuli, e.g., cells treated with LPS or TNF. The in vivo administration of nonblocking, nonagonistic TR75-89 antibody reportedly results in the linear accumulation of shed, soluble forms of p75 TNFR in the circulation. The TR75-89 antibody does not recognize the 55 kDa (p55) Type I TNFR (aka, CD120a). TR75-89 does not block the binding and does not neutralize the bioactivity of the TNF ligand on L929 target cell populations that express p75 (and p55) TNFR. The immunogen used to generate the TR75-89 hybridoma was a purified, soluble extracellular domain of the mouse Type II TNFR.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD120b
Applications Flow Cytometry
Classification Monoclonal
Clone TR75-89
Concentration 0.2mg/mL
Conjugate Brilliant Violet 421
Description Tnfrsf1b; Tnfr2; TNF-R2; TNFR p75; TNFRII; TNF-R-II; TNFBR
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Armenian Hamster
Immunogen Mouse Type II TNFR
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 λ3
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.