Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD120b Rat anti-Human, PE, Clone: HTNFR-M1, BD

Catalog No. BDB552418 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB552418 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB552418 Supplier BD Biosciences Supplier No. 552418
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The hTNFR-M1 antibody reacts with the extracellular domain of the 75 kDa transmembrane receptor for the human cytokines, tumor necrosis factor (TNF or TNF-α) and lymphotoxin-alpha (LT-α3, aka, lymphotoxin or TNF-β). This receptor is referred to as the p75 or Type II Tumor Necrosis Factor Receptor (TNFRII) [aka, CD120b]. Human TNFRII proteins are expressed by hematopoietic cells including macrophages, neutrophils, lymphocytes, thymocytes and mast cells. TNFRII is expressed by a variety of other cell types including endothelial cells, cardiac myocytes and prostate cells. Naive B cells express very low or undetectable levels of TNFRII whereas mature erythrocytes and platelets are uniformly negative for TNFRII expression. The immunogen used to generate the hTNFR-M1 hybridoma was COS- expressed recombinant human TNFRII.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD120b
Applications Flow Cytometry
Classification Monoclonal
Clone HTNFR-M1
Conjugate PE
Description TNFRSF1B; p75 TNFR; p75 TNF Receptor; TNFR80; TNF-RII; TNF-R2
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Rat
Immunogen COS-expressed recombinant human TNFRII
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.