Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD121a Rat anti-Mouse, Alexa Fluor 647, Clone: 35F5, BD

Catalog No. BDB563629 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB563629 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB563629 Supplier BD Biosciences Supplier No. 563629

Rat Monoclonal Antibody

The 35F5 monoclonal antibody specifically binds to the Type I Interleukin-1 Receptor (IL-1 RI) which is also known as CD121a. CD121a is expressed by T-cell, fibroblast, and epithelial cell lines such as EL-4 and 3T3. Expression of the 35F5 epitope on normal mouse thymocytes and splenocytes is undetectable by flow cytometry. CD121a is an 80 kDa type-I transmembrane glycoprotein, a member of the Ig superfamily that mediates the inflammatory responses of leucocytes to IL-1. The 35F5 antibody is capable of blocking the binding of IL-1α, IL-1β, and IL-1 receptor antagonist to the Type I IL-1 Receptor. The 35F5 mAb is highly specific for the Type I IL-1 Receptor and does not react with the Type II IL-1 Receptor (CD121b) which is present on mouse macrophage and B-cell lines and polymorphonuclear leukocytes (PMN) and PMN progenitors.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD121a
Applications Flow Cytometry
Classification Monoclonal
Clone 35F5
Concentration 0.2mg/mL
Conjugate Alexa Fluor 647
Description Il1r1; IL-1 Receptor Type I/p80; IL-1 R alpha; IL-1 Rα; IL-1 RI
Host Species Rat
Immunogen IL-1 receptors from the C57BL/6 mouse T lymphoma EL4.IL2
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.